Invitrogen
Manufacturer Code:PA5114340
Catalog # PIPA5114340
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide within A. thaliana PAS2 aa 180-221. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxyacyl-CoA dehydratase PASTICCINO 2; AtPAS2; PEP; Protein PEPINO; Protein tyrosine phosphatase-like protein; Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase PASTICCINO 2
Gene Aliases: AT5G10480; F12B17.170; F12B17_170; PAS2; PASTICCINO 2; PEP; PEPINO
UniProt ID: (Arabidopsis) Q8VZB2
Entrez Gene ID: (Arabidopsis) 830912
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.