Invitrogen
Manufacturer Code:PA529309
Catalog # PIPA529309
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 982 and 1227 of Human PASK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAS domain-containing serine/threonine-protein kinase; PAS-kinase; paskin; PASKIN STK37 PAS domain-containing serine/threonine-protein kinase per-arnt-sim (PAS) domain kinase; per-arnt-sim (PAS) domain kinase
Gene Aliases: KIAA0135; mKIAA0135; PASK; PASKIN; STK37
UniProt ID: (Human) Q96RG2, (Mouse) Q8CEE6
Entrez Gene ID: (Human) 23178, (Mouse) 269224
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.