Invitrogen
Manufacturer Code:PA552800
Catalog # PIPA552800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human PASD1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cancer/testis antigen 63; Circadian clock protein PASD1; CT63; CT63 OXTES1 PAS domain-containing protein 1 cancer/testis antigen 63; OX-TES-1; PAS domain-containing protein 1
Gene Aliases: CT63; OXTES1; PASD1
UniProt ID: (Human) Q8IV76
Entrez Gene ID: (Human) 139135
Molecular Function: basic helix-loop-helix transcription factor transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.