Invitrogen
Manufacturer Code:PA595781
Catalog # PIPA595781
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit funsion protein containing a sequence corresponding to amino acids 30-220 of human PARS2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Probable proline--tRNA ligase, mitochondrial; probable prolyl-tRNA synthetase, mitochondrial; proline--tRNA ligase; Prolyl-tRNA synthetase; prolyl-tRNA synthetase (mitochondrial)(putative); ProRS
Gene Aliases: Pars2; RGD1305345
UniProt ID: (Rat) Q5M7W7
Entrez Gene ID: (Rat) 313429
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.