Novus Biologicals
Manufacturer Code:NBP29459520UL
Catalog # NB023715
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 30-220 of human PARS2 (NP_689481.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp727A071 EC 6.1.1.15 MGC14416 MGC19467 MT-PRORS probable prolyl-tRNA synthetase mitochondrial proline tRNA ligase 2 mitochondrial (putative) Proline--tRNA ligase prolyl-tRNA synthetase (mitochondrial)(putative) prolyl-tRNA synthetase 2 mitochondrial (putative) prolyl-tRNA synthetase mitochondrial (putative) proRS; Probable proline--tRNA ligase, mitochondrial; proline tRNA ligase 2, mitochondrial (putative); Prolyl-tRNA synthetase; ProRS
Gene Aliases: MT-PRORS; PARS2; proRS
UniProt ID: (Human) Q7L3T8
Entrez Gene ID: (Human) 25973
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.