Novus Biologicals
Manufacturer Code:NBP155363
Catalog # NBP155363
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to TIPARP(TCDD-inducible poly(ADP-ribose) polymerase) The peptide sequence was selected from the N terminal of TIPARP. Peptide sequence LKTCFKKKDQKRLGTGTLRSLRPILNTLLESGSLDGVFRSRNQSTDENSL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 14; ARTD14; DDF1 DKFZP434J214 DKFZp686N0351 DKFZp686P1838 EC 2.4.2.30 FLJ40466 PARP-1 PARP-7 PARP7DKFZp434J214 pART14 Poly [ADP-ribose] polymerase 7 RM1 TCDD-inducible poly [ADP-ribose] polymerase TCDD-inducible poly(ADP-ribose) polymerase; PARP-7; Poly [ADP-ribose] polymerase 7; Protein mono-ADP-ribosyltransferase TIPARP; TCDD-inducible poly [ADP-ribose] polymerase; TCDD-inducible poly(ADP-ribose) polymerase
Gene Aliases: ARTD14; PARP7; pART14; TIPARP
UniProt ID: (Human) Q86VP4
Entrez Gene ID: (Human) 25976
Molecular Function: nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.