Invitrogen
Manufacturer Code:PA5112640
Catalog # PIPA5112640
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Poly [ADP-ribose] polymerase 3 protein (294-533AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A DP-ribosyltransferase (NAD+ poly (ADP-ribose polymerase)-like 3 A930002C11Rik ADP-ribosyltransferase (NAD+ poly (ADP-ribose polymerase)-like 3 ADP-ribosyltransferase (NAD+ poly (ADP-ribose polymerase)-like 3 ADP-ribosyltransferase (NAD+ poly (ADP-ribose) polymerase)-like 2 ADP-ribosyltransferase (NAD+ poly (ADP-ribose) polymerase)-like 3 ADP-ribosyltransferase diphtheria toxin-like 3 ADPRT3 ADPRT-3 ADPRTL2 ADPRTL3 ARTD3 AW990611 DNA ADP-ribosyltransferase PARP3 IRT1 NAD(+) ADP-ribosyltransferase 3 NAD+ ADP-ribosyltransferase 3 pADPRT-3 Parp3 PARP-3 poly (ADP-ribose) polymerase family member 3 poly (ADP-ribosyl) transferase-like 3 Poly [ADP-ribose] polymerase 3 poly(ADP-ribose) polymerase family member 3 poly(ADP-ribose) synthetase-3 Poly[ADP-ribose] synthase 3 poly[ADP-ribose] synthetase 3 Poly[ADP-ribose] synthetase-3 Protein mono-ADP-ribosyltransferase PARP3; A DP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+, poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose polymerase)-like 3; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase)-like 3; ADP-ribosyltransferase diphtheria toxin-like 3; ADPRT-3; ARTD3; DNA ADP-ribosyltransferase PARP3; IRT1; NAD(+) ADP-ribosyltransferase 3; NAD+ ADP-ribosyltransferase 3; pADPRT-3; PARP-3; poly (ADP-ribose) polymerase family, member 3; Poly [ADP-ribose] polymerase 3; poly(ADP-ribose) synthetase-3; Poly[ADP-ribose] synthase 3; poly[ADP-ribose] synthetase 3; Poly[ADP-ribose] synthetase-3; Protein mono-ADP-ribosyltransferase PARP3
Gene Aliases: A930002C11Rik; ADPRT3; ADPRTL2; ADPRTL3; ARTD3; AW990611; IRT1; PADPRT-3; PARP-3; PARP3
UniProt ID: (Human) Q9Y6F1, (Mouse) Q3ULW8
Entrez Gene ID: (Human) 10039, (Mouse) 235587
Molecular Function: DNA binding protein DNA ligase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.