Invitrogen
Manufacturer Code:MA534728
Catalog # PIMA534728
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JG34-56 |
Immunogen | Recombinant protein within the C-terminus of human PARP2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles store in dark |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase diphtheria toxin-like 2; ADP-ribosyltransferase diphtheria toxin-like 2 ADPRT2 ADPRT-2 ADPRTL2 ADPRTL3 ARTD2 Aspartl2 C78626 DNA ADP-ribosyltransferase PARP2 HGNC:272 hPARP-2 NAD(+) ADP-ribosyltransferase 2 pADPRT-2 Parp2 PARP-2 Poly [ADP-ribose] polymerase 2 Poly[ADP-ribose] synthase 2 Protein poly-ADP-ribosyltransferase PARP2; ADPRT-2; ARTD2; DNA ADP-ribosyltransferase PARP2; hPARP-2; NAD(+) ADP-ribosyltransferase 2; pADPRT-2; PARP-2; poly (ADP-ribose) polymerase 2; poly (ADP-ribose) polymerase family, member 2; poly (ADP-ribosyl) transferase-like 2; Poly [ADP-ribose] polymerase 2; Poly[ADP-ribose] synthase 2; poly[ADP-ribose] synthetase 2; Protein poly-ADP-ribosyltransferase PARP2
Gene Aliases: ADPRT2; ADPRTL2; ADPRTL3; ARTD2; pADPRT-2; PARP-2; PARP2
UniProt ID: (Human) Q9UGN5
Entrez Gene ID: (Human) 10038
Molecular Function: DNA binding protein DNA ligase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.