Invitrogen
Manufacturer Code:MA538149
Catalog # PIMA538149
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC2099 |
Immunogen | A synthesized peptide derived from human PARP2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase (NAD+ poly(ADP-ribose) polymerase)-like 2 ADP-ribosyltransferase (NAD+ poly (ADP-ribose) polymerase) 2 ADP-ribosyltransferase (NAD+ poly(ADP-ribose) polymerase)-like 2 ADP-ribosyltransferase diphtheria toxin-like 2 ADPRT2 ADPRT-2 Adprtl2 ADPRTL3 ARTD2 Aspartl2 C78626 DNA ADP-ribosyltransferase PARP2 HGNC:272 hPARP-2 mPARP-2 NAD(+) ADP-ribosyltransferase 2 pADPRT-2 parp 2 Parp2 PARP-2 poly (ADP-ribose) polymerase 2 poly (ADP-ribose) polymerase family member 2 poly (ADP-ribosyl) transferase-like 2 poly [ADP-ribose] polymerase 2 poly(ADP-ribose) polymerase 2 poly[ADP-ribose] synthase 2 poly[ADP-ribose] synthetase 2 Protein poly-ADP-ribosyltransferase PARP2; ADP-ribosyltransferase (NAD+, poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase) 2; ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase diphtheria toxin-like 2; ADPRT-2; ARTD2; DNA ADP-ribosyltransferase PARP2; hPARP-2; mPARP-2; NAD(+) ADP-ribosyltransferase 2; pADPRT-2; PARP-2; poly (ADP-ribose) polymerase 2; poly (ADP-ribose) polymerase family, member 2; poly (ADP-ribosyl) transferase-like 2; Poly [ADP-ribose] polymerase 2; Poly[ADP-ribose] synthase 2; poly[ADP-ribose] synthetase 2; Protein poly-ADP-ribosyltransferase PARP2
Gene Aliases: ADPRT2; ADPRTL2; ADPRTL3; ARTD2; Aspartl2; C78626; pADPRT-2; PARP-2; PARP2
UniProt ID: (Human) Q9UGN5, (Mouse) O88554
Entrez Gene ID: (Human) 10038, (Rat) 290027, (Mouse) 11546
Molecular Function:
DNA binding protein
DNA ligase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.