Novus Biologicals
Manufacturer Code:NBP15468020UL
Catalog # NBP15468020
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to PARP2 (poly (ADP-ribose) polymerase family member 2) The peptide sequence was selected from the middle region of PARP2. Peptide sequence LLDLFEVEKDGEKEAFREDLHNRMLLWHGSRMSNWVGILSHGLRIAHPEA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase (NAD+ poly(ADP-ribose) polymerase)-like 2 ADPRT-2 ADPRT2pADPRT-2 ADPRTL2EC 2.4.2.30 ADPRTL3 hPARP-2 NAD(+) ADP-ribosyltransferase 2 PARP-2 poly (ADP-ribose) polymerase 2 poly (ADP-ribose) polymerase family member 2 poly (ADP-ribosyl) transferase-like 2 poly [ADP-ribose] polymerase 2 poly(ADP-ribose) synthetase Poly[ADP-ribose] synthase 2 poly[ADP-ribose] synthetase 2; ADP-ribosyltransferase (NAD+; poly(ADP-ribose) polymerase)-like 2; ADP-ribosyltransferase diphtheria toxin-like 2; ADPRT-2; ARTD2; DNA ADP-ribosyltransferase PARP2; hPARP-2; NAD(+) ADP-ribosyltransferase 2; pADPRT-2; PARP-2; poly (ADP-ribose) polymerase 2; poly (ADP-ribose) polymerase family, member 2; poly (ADP-ribosyl) transferase-like 2; Poly [ADP-ribose] polymerase 2; Poly[ADP-ribose] synthase 2; poly[ADP-ribose] synthetase 2; Protein poly-ADP-ribosyltransferase PARP2
Gene Aliases: ADPRT2; ADPRTL2; ADPRTL3; ARTD2; pADPRT-2; PARP-2; PARP2
UniProt ID: (Human) Q9UGN5
Entrez Gene ID: (Human) 10038
Molecular Function:
DNA binding protein
DNA ligase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.