OriGene
Manufacturer Code:TA500453
Catalog # 50-167-1588
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4E2 |
Immunogen | Full-length protein expressed in 293T cell transfected with human PARP16 expression vector. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase diphtheria toxin-like 15; ARTD15 C15orf30 pART15; PARP-16; poly (ADP-ribose) polymerase family, member 16; Poly [ADP-ribose] polymerase 16; Protein mono-ADP-ribosyltransferase PARP16
Gene Aliases: ARTD15; C15orf30; PARP16; pART15
UniProt ID: (Human) Q8N5Y8
Entrez Gene ID: (Human) 54956
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.