Invitrogen
Manufacturer Code:PA5114686
Catalog # PIPA5114686
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The antiserum was produced against synthesized peptide derived from human PARP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 5830444G22Rik ADP-ribosyltransferase (NAD+ ADP-ribosyltransferase (NAD+) poly (ADP-ribose) polymerase) ADP-ribosyltransferase (NAD+ poly (ADP-ribose) polymerase) 1 ADP-ribosyltransferase (NAD+ poly (ADP-ribose) polymerase) ADP-ribosyltransferase (NAD+ poly (ADP-ribose) polymerase) 1 ADP-ribosyltransferase 1 ADP-ribosyltransferase diphtheria toxin-like 1 ADP-ribosyltransferase NAD(+) Adprp Adprt ADPRT 1 ADPRT1 AI893648 ARTD1 C80510 DNA ADP-ribosyltransferase PARP1 msPARP NAD(+) ADP-ribosyltransferase 1 pADPRT-1 PARP Parp1 parp-1 poly (ADP-ribose) polymerase 1 poly (ADP-ribose) polymerase family member 1 poly (ADP-ribose) polymerase) poly [ADP-ribose] polymerase 1 poly(ADP-ribose) polymerase poly(ADP-ribose) polymerase 1 poly(ADP-ribose) polymerase PARP-1 poly(ADP-ribose) synthetase poly(ADP-ribosyl)transferase poly[ADP-ribose] synthase 1 poly[ADP-ribose] synthetase 1 PPOL Protein poly-ADP-ribosyltransferase PARP1 sPARP-1 synthetase fragment (AA 67) Unknown (protein for IMAGE:8023736) unnamed protein product; ADP-ribosyltransferase (NAD+; ADP-ribosyltransferase (NAD+, poly (ADP-ribose) polymerase) 1; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase); ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase) 1; ADP-ribosyltransferase 1; ADP-ribosyltransferase diphtheria toxin-like 1; ADP-ribosyltransferase NAD(+); ADPRT 1; ARTD1; DNA ADP-ribosyltransferase PARP1; msPARP; NAD(+) ADP-ribosyltransferase 1; PARP-1; poly (ADP-ribose) polymerase 1; poly (ADP-ribose) polymerase family, member 1; poly (ADP-ribose) polymerase); Poly [ADP-ribose] polymerase 1; poly(ADP-ribose) polymerase; poly(ADP-ribose) polymerase PARP-1; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase; Poly[ADP-ribose] synthase 1; poly[ADP-ribose] synthetase 1; Protein poly-ADP-ribosyltransferase PARP1
Gene Aliases: 5830444G22Rik; Adprp; ADPRT; ADPRT 1; ADPRT1; AI893648; ARTD1; C80510; pADPRT-1; PARP; PARP-1; PARP1; PPOL; sPARP-1
UniProt ID: (Human) P09874, (Rat) P27008
Entrez Gene ID: (Human) 142, (Rat) 25591, (Mouse) 11545
Molecular Function: DNA binding protein DNA ligase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.