Novus Biologicals
Manufacturer Code:NB10056599
Catalog # NB10056599
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 194C1439 |
Immunogen | This antibody was developed by immunizing mice with a synthetic peptide containing amino acids near 214/215-cleavage site of human PARP. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ADP-ribosyltransferase (NAD+ poly (ADP-ribose) polymerase) ADPRT 1 ADPRTADP-ribosyltransferase NAD(+) cleaved PARP cleaved PARP 1 cleaved PARP apoptosis cleaved PARP western blot cleaved PARP-1 EC 2.4.2 EC 2.4.2.30 NAD(+) ADP-ribosyltransferase 1 PARP apoptosis PARP-1 PARPADPRT1 poly (ADP-ribose) polymerase 1 poly (ADP-ribose) polymerase family member 1 poly [ADP-ribose] polymerase 1 poly(ADP-ribose) polymerase poly(ADP-ribose) synthetase poly(ADP-ribosyl)transferase Poly[ADP-ribose] synthase 1 PPOLpADPRT-1; ADP-ribosyltransferase (NAD+; poly (ADP-ribose) polymerase); ADP-ribosyltransferase diphtheria toxin-like 1; ADP-ribosyltransferase NAD(+); ADPRT 1; ARTD1; DNA ADP-ribosyltransferase PARP1; NAD(+) ADP-ribosyltransferase 1; PARP-1; poly (ADP-ribose) polymerase 1; poly (ADP-ribose) polymerase family, member 1; Poly [ADP-ribose] polymerase 1; poly(ADP-ribose) polymerase; poly(ADP-ribose) synthetase; poly(ADP-ribosyl)transferase; Poly[ADP-ribose] synthase 1; Protein poly-ADP-ribosyltransferase PARP1
Gene Aliases: ADPRT; ADPRT 1; ADPRT1; ARTD1; pADPRT-1; PARP; PARP-1; PARP1; PPOL
UniProt ID: (Human) P09874
Entrez Gene ID: (Human) 142
Molecular Function: DNA binding protein DNA ligase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.