Proteintech
Manufacturer Code:137991AP150UL
Catalog # 50-172-9015
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PARN Fusion Protein Ag4774 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DAN Deadenylating nuclease Deadenylation nuclease PARN; Deadenylating nuclease; Deadenylation nuclease; poly(A)-specific ribonuclease (deadenylation nuclease); Poly(A)-specific ribonuclease PARN; Polyadenylate-specific ribonuclease
Gene Aliases: 1200003I18Rik; DAN; DKCB6; PARN; PFBMFT4; RGD1565449
UniProt ID: (Human) O95453, (Mouse) Q8VDG3
Entrez Gene ID: (Human) 5073, (Mouse) 74108, (Rat) 360464
Molecular Function:
RNA binding protein
mRNA polyadenylation factor
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.