Invitrogen
Manufacturer Code:PA572553
Catalog # PIPA572553
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 1329-1356 amino acids from the C-terminal region of human PARD3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASIP; atypical PKC isotype-specific interacting protein; Atypical PKC isotype-specific-interacting protein; bazooka; CTCL tumor antigen se2-5; Ephrin-interacting protein; PAR-3; par-3 (partitioning defective 3) homolog; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3-alpha; Partitioning defective 3 homolog; PHIP; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1 ASIP Baz Bazooka FLJ21015 PAR3 PAR3alpha PARD3A SE2-5L16 SE2-5LT1 SE2-5T2 CTCL tumor antigen se2-5 PAR3-alpha PARD-3 atypical PKC isotype-specific interacting protein atypical PKC isotype-specific-interacting protein partitioning defective 3 homolog
Gene Aliases: AA960621; AI256638; ASIP; Baz; D8Ertd580e; Par-3; PAR3; PAR3A; PAR3alpha; PARD-3; PARD3; PARD3A; Phip; PPP1R118; SE2-5L16; SE2-5LT1; SE2-5T2
UniProt ID: (Human) Q8TEW0, (Mouse) Q99NH2
Entrez Gene ID: (Human) 56288, (Mouse) 93742
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.