Invitrogen
Manufacturer Code:PA519791
Catalog # PIPA519791
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide conjugated to KLH derived from within residues 1300 to the C-terminus of Human PARD3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C or -80°C if preferred |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ASBP; ASIP; atypical PKC isotype-specific interacting protein; Atypical PKC isotype-specific-interacting protein; atypical PKC-specific binding protein; Atypical PKC-specific-binding protein; bazooka; CTCL tumor antigen se2-5; PAR-3; par-3 (partitioning defective 3) homolog; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3-alpha; PARD-3; Partitioning defective 3 homolog; partitioning-defective 3 homolog; protein phosphatase 1, regulatory subunit 118; RP11-406D17.1 ASIP Baz Bazooka FLJ21015 PAR3 PAR3alpha PARD3A SE2-5L16 SE2-5LT1 SE2-5T2 CTCL tumor antigen se2-5 PAR3-alpha PARD-3 atypical PKC isotype-specific interacting protein atypical PKC isotype-specific-interacting protein partitioning defective 3 homolog; three-PDZ containing protein similar to C. elegans PAR3 (partitioning defect)
Gene Aliases: ASIP; Baz; PAR3; PAR3A; PAR3alpha; PARD-3; PARD3; PARD3A; PPP1R118; SE2-5L16; SE2-5LT1; SE2-5T2
UniProt ID: (Human) Q8TEW0, (Rat) Q9Z340
Entrez Gene ID: (Human) 56288, (Rat) 81918
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.