Invitrogen
Manufacturer Code:PA5106519
Catalog # PIPA5106519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human PARD3 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AA960621 AI256638 ASBP ASIP atypical PKC isotype-specific interacting protein Atypical PKC isotype-specific-interacting protein atypical PKC-specific binding protein atypical PKC-specific-binding protein Baz bazooka CTCL tumor antigen se2-5 D8Ertd580e Ephrin-interacting protein FLJ21015 PAR3 PAR-3 par-3 (partitioning defective 3) homolog par-3 family cell polarity regulator par-3 family cell polarity regulator alpha par-3 partitioning defective 3 homolog PAR3A PAR3alpha PAR3-alpha Pard3 PARD-3 Pard3a Partitioning defective 3 homolog partitioning-defective 3 homolog PHIP PPP1R118 protein phosphatase 1 regulatory subunit 118 RP11-406D17.1 SE2-5L16 SE2-5LT1 SE2-5T2 three-PDZ containing protein similar to C. elegans PAR3 (partitioning defect); ASBP; ASIP; atypical PKC isotype-specific interacting protein; Atypical PKC isotype-specific-interacting protein; atypical PKC-specific binding protein; Atypical PKC-specific-binding protein; bazooka; CTCL tumor antigen se2-5; Ephrin-interacting protein; PAR-3; par-3 (partitioning defective 3) homolog; par-3 family cell polarity regulator alpha; par-3 partitioning defective 3 homolog; PAR3-alpha; PARD-3; Partitioning defective 3 homolog; partitioning-defective 3 homolog; PHIP; protein phosphatase 1, regulatory subunit 118; three-PDZ containing protein similar to C. elegans PAR3 (partitioning defect)
Gene Aliases: AA960621; AI256638; ASIP; Baz; D8Ertd580e; Par-3; PAR3; PAR3A; PAR3alpha; PARD-3; PARD3; PARD3A; Phip; PPP1R118; SE2-5L16; SE2-5LT1; SE2-5T2
UniProt ID: (Human) Q8TEW0, (Mouse) Q99NH2, (Rat) Q9Z340
Entrez Gene ID: (Human) 56288, (Mouse) 93742, (Rat) 81918
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.