Invitrogen
Manufacturer Code:PA518233
Catalog # PIPA518233
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide sequence (GSRIRGDGSGFSL) corresponding to the C-terminus amino acids of PARD6A |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: PAR-6; PAR-6 alpha; par-6 partitioning defective 6 homolog alpha; PAR6C; PARD6A PAR6alpha PAR-6 TAX40 PAR-6A TIP-40 par-6 partitioning defective 6 homolog alpha (C.elegans) Tax interaction protein 40 par-6 (partitioning defective 6 C.elegans) homolog alpha PAR6 PAR6C Tax-interacting protein 40 par-6 partitioning defective 6 homolog alpha partitioning defective-6 homolog alpha partitioning-defective protein 6; Partitioning defective 6 homolog alpha; partitioning defective-6 homolog alpha; partitioning-defective protein 6; Tax interaction protein 40; Tax-interacting protein 40; TIP-40
Gene Aliases: PAR-6A; PAR6; PAR6A; PAR6alpha; PAR6C; PARD6A; TAX40; TIP-40
UniProt ID: (Human) Q9NPB6
Entrez Gene ID: (Human) 50855
Molecular Function:
cell junction protein
tight junction
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.