Invitrogen
Manufacturer Code:MA538130
Catalog # PIMA538130
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1980 |
Immunogen | A synthesized peptide derived from human PAR4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Recombinant Monoclonal Antibody
For Research Use Only
Protein Aliases: 2310001G03Rik PAR4 Par-4 par-4 induced by effectors of apoptosis Pawr PRKC apoptosis WT1 regulator protein PRKC apoptosis WT1 regulator pro-apoptotic WT1 regulator prostate apoptosis response 4 prostate apoptosis response 4 protein Prostate apoptosis response protein 4 prostate apoptosis response protein PAR-4 prostate apoptosis response-4 transcriptional repressor PAR4 Transcriptional repressor Par-4-like protein PAWR WT1-interacting protein; Par-4; PRKC apoptosis WT1 regulator protein; PRKC, apoptosis, WT1, regulator; Prostate apoptosis response 4 protein; prostate apoptosis response protein 4; prostate apoptosis response protein PAR-4; prostate apoptosis response-4; transcriptional repressor PAR4; WT1-interacting protein
Gene Aliases: 2310001G03Rik; Par-4; PAR4; PAWR
UniProt ID: (Human) Q96IZ0, (Mouse) Q925B0
Entrez Gene ID: (Human) 5074, (Mouse) 114774
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.