R&D Systems
Manufacturer Code:FAB3949A
Catalog # FAB3949A
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 344222 |
Immunogen | NS0 mouse myeloma cell line transfected with human PAR2 Accession # P55085 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CFIIRL1 coagulation factor II (thrombin) receptor-like 1 Coagulation factor II receptor-like 1 F2RL1 GPCR11 GPR11protease-activated receptor 2 G-protein coupled receptor 11 PAR-2 PAR2G protein-coupled receptor-11 proteinase-activated receptor 2 proteinase-activated receptor-2 Thrombin receptor-like 1; coagulation factor II (thrombin) receptor-like 1; Coagulation factor II receptor-like 1; G-protein coupled receptor 11; PAR-2; protease-activated receptor 2; Proteinase-activated receptor 2; Proteinase-activated receptor 2, alternate cleaved 1; Proteinase-activated receptor 2, alternate cleaved 2; Thrombin receptor-like 1
Gene Aliases: F2RL1; GPR11; PAR2
UniProt ID: (Human) P55085
Entrez Gene ID: (Human) 2150
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.