Proteintech
Manufacturer Code:121601AP150UL
Catalog # 50-172-7757
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PAR2 Fusion Protein Ag2801 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: coagulation factor II (thrombin) receptor-like 1; Coagulation factor II receptor-like 1; F2RL1 G protein coupled receptor 11 GPR11 PAR 2 PAR2 Thrombin receptor like 1; G-protein coupled receptor 11; PAR-2; protease-activated receptor 2; Protease-activated receptor-2; Proteinase-activated receptor 2; Proteinase-activated receptor 2, alternate cleaved 1; Proteinase-activated receptor 2, alternate cleaved 2; proteinase-activated receptor-2; Proteinase-activated receptor-2 G protein-coupled receptor 11; Proteinase-activated receptor-2, G protein-coupled receptor 11; Thrombin receptor-like 1
Gene Aliases: F2RL1; Gpcr11; GPR11; PAR-2; PAR2
UniProt ID: (Human) P55085, (Mouse) P55086, (Rat) Q63645
Entrez Gene ID: (Human) 2150, (Mouse) 14063, (Rat) 116677
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.