Novus Biologicals
Manufacturer Code:NBP171770SS
Catalog # NBP171770SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | N2-11 |
Immunogen | The oligopeptide CNATLDPRSFLL from human Thrombin Receptor. [Swiss-Prot# P25116] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CF2R CF2RHTR coagulation factor II (thrombin) receptor Coagulation factor II receptor HTR PAR1 PAR1PAR-1protease-activated receptor 1 Protease-Activated Receptor 1 Thrombin receptor TRproteinase-activated receptor 1; coagulation factor II (thrombin) receptor; Coagulation factor II receptor; PAR-1; protease activated receptor 1; protease-activated receptor 1; Proteinase-activated receptor 1; Thrombin receptor
Gene Aliases: CF2R; F2R; HTR; PAR-1; PAR1; TR
UniProt ID: (Human) P25116
Entrez Gene ID: (Human) 2149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.