R&D Systems
Manufacturer Code:MAB3855
Catalog # MAB3855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 731115 |
Immunogen | <EM>E. coli</EM>-derived recombinant human PAR1 Arg27-Thr102 Ser375-Thr425 Accession # P25116 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: CF2R CF2RHTR coagulation factor II (thrombin) receptor Coagulation factor II receptor F2R HTR PAR1 PAR1PAR-1protease-activated receptor 1 Protease-Activated Receptor 1 Thrombin receptor ThrR TRGPC TRproteinase-activated receptor 1; coagulation factor II (thrombin) receptor; Coagulation factor II receptor; PAR-1; protease activated receptor 1; protease-activated receptor 1; Proteinase-activated receptor 1; Thrombin receptor
Gene Aliases: CF2R; F2R; HTR; PAR-1; PAR1; TR
UniProt ID: (Human) P25116
Entrez Gene ID: (Human) 2149
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.