Invitrogen
Manufacturer Code:PA5116040
Catalog # PIPA5116040
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human Thrombin Receptor corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI482343 C HTR Cf2r coagulation factor II (thrombin) receptor coagulation factor II receptor coagulation factor II thrombin receptor F2R HTR PAR1 PAR-1 protease activated receptor 1 protease-activated receptor 1 proteinase-activated receptor 1 Thrombin receptor ThrR TR TRGPC; coagulation factor II (thrombin) receptor; Coagulation factor II receptor; PAR-1; protease activated receptor 1; protease-activated receptor 1; Proteinase-activated receptor 1; Thrombin receptor
Gene Aliases: AI482343; CF2R; F2R; HTR; PAR-1; PAR1; ThrR; TR; TRGPC
UniProt ID: (Human) P25116, (Mouse) P30558
Entrez Gene ID: (Human) 2149, (Mouse) 14062, (Rat) 25439
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.