OriGene
Manufacturer Code:CF807111
Catalog # 50-166-8123
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1E12 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-247 of human PAPSS2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 3'-phosphoadenosine-5'-phosphosulfate synthase; 3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2; adenosine 5'-phosphosulfate kinase; Adenosine-5'-phosphosulfate 3'-phosphotransferase; Adenylyl-sulfate kinase; Adenylylsulfate 3'-phosphotransferase; APS kinase; ATP sulfurylase/adenosine 5'-phosphosulfate kinase; ATP sulfurylase/APS kinase 2; ATP-sulfurylase; ATPSK2 BCYM4 SK2; Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2; bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2; PAPS synthase 2; PAPS synthetase 2; SAT; SK 2; Sulfate adenylate transferase; Sulfate adenylyltransferase; Sulfurylase kinase 2
Gene Aliases: ATPSK2; BCYM4; PAPSS2; SK2
UniProt ID: (Human) O95340
Entrez Gene ID: (Human) 9060
Molecular Function: kinase nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.