Invitrogen
Manufacturer Code:PA5111671
Catalog # PIPA5111671
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Protein corresonding to Human PAPSS2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1810018P12Rik 3&prime-phosphoadenosine-5&prime-phosphosulfate synthase Adenosine-5&prime-phosphosulfate 3&prime-phosphotransferase Adenylylsulfate 3&prime-phosphotransferase Adenylyl-sulfate kinase AI159688 APS kinase ATPSK2 AtpsU2 ATP-sulfurylase BCYM4 Bifunctional 3&prime-phosphoadenosine 5&prime-phosphosulfate synthase 2 bm PAPS synthase 2 PAPS synthethase 2 PAPSS 2 Papss2 SAT SK 2 SK2 Sulfate adenylate transferase Sulfate adenylyltransferase Sulfurylase kinase 2; 3'-phosphoadenosine-5'-phosphosulfate synthase; 3-prime-phosphoadenosine 5-prime-phosphosulfate synthase 2; adenosine 5'-phosphosulfate kinase; Adenosine-5'-phosphosulfate 3'-phosphotransferase; Adenylyl-sulfate kinase; Adenylylsulfate 3'-phosphotransferase; APS kinase; ATP sulfurylase/adenosine 5'-phosphosulfate kinase; ATP sulfurylase/APS kinase 2; ATP-sulfurylase; Bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthase 2; bifunctional 3'-phosphoadenosine 5'-phosphosulfate synthethase 2; PAPS synthase 2; PAPS synthetase 2; SAT; SK 2; Sulfate adenylate transferase; Sulfate adenylyltransferase; Sulfurylase kinase 2
Gene Aliases: ATPSK2; BCYM4; PAPSS2; SK2
UniProt ID: (Human) O95340
Entrez Gene ID: (Human) 9060
Molecular Function: kinase nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.