Proteintech
Manufacturer Code:242841AP150UL
Catalog # 50-173-4276
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PAPOLG Fusion Protein Ag16208 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Neo PAP Neo poly(A) polymerase PAP gamma PAP2 PAPG PAPOLG poly(A) polymerase gamma SRP RNA 3 adenylating enzyme; Neo-PAP; Neo-poly(A) polymerase; nuclear poly(A) polymerase gamma; PAP-gamma; Poly(A) polymerase gamma; Polynucleotide adenylyltransferase gamma; Signal recognition particle RNA-adenylating enzyme; SRP RNA 3' adenylating enzyme/pap2; SRP RNA 3'-adenylating enzyme; SRP RNA-adenylating enzyme
Gene Aliases: PAP2; PAPG; PAPOLG
UniProt ID: (Human) Q9BWT3
Entrez Gene ID: (Human) 64895
Molecular Function:
RNA binding protein
mRNA polyadenylation factor
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.