Proteintech
Manufacturer Code:128211AP150UL
Catalog # 50-172-8275
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PAPOLB Fusion Protein Ag3487 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAP beta PAPOLB PAPT Poly(A) polymerase beta TPAP; PAP-beta; Poly(A) polymerase beta; poly(A) polymerase beta (testis specific); Polynucleotide adenylyltransferase beta; Testis-specific poly(A) polymerase
Gene Aliases: PAP; Paplob; Papola-ps; PAPOLB; PAPT; Plap-ps; TPAP
UniProt ID: (Human) Q9NRJ5, (Mouse) Q9WVP6
Entrez Gene ID: (Human) 56903, (Rat) 304300, (Mouse) 56522
Molecular Function:
RNA binding protein
mRNA polyadenylation factor
mRNA processing factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.