Invitrogen
Manufacturer Code:PA572659
Catalog # PIPA572659
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 378-413 amino acids from the C-terminal region of human (DANRE) papl |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Acid phosphatase type 7; iron/zinc purple acid phosphatase-like protein; Iron/zinc purple acid phosphatase-like protein papl; Purple acid phosphatase long form
Gene Aliases: acp7; papl; zgc:162913
UniProt ID: (Zebrafish) A5D6U8
Entrez Gene ID: (Zebrafish) 571830
Molecular Function: hydrolase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.