Invitrogen
Manufacturer Code:PA570001
Catalog # PIPA570001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of rat PAOX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: exo-N4-amino; PAO peroxisomal N(1)-acetyl-spermine/spermidine oxidase peroxisomal N1-acetyl-spermine/spermidine oxidase; Peroxisomal N(1)-acetyl-spermine/spermidine oxidase; peroxisomal N1-acetyl-spermine/spermidine oxidase; Polyamine oxidase; putative GTP-binding protein
Gene Aliases: 2410012F02Rik; AI118225; PAO; PAOX; UNQ1923/PRO4398
UniProt ID: (Human) Q6QHF9, (Bovine) Q865R1, (Mouse) Q8C0L6
Entrez Gene ID: (Horse) 100629322, (Human) 196743, (Bovine) 282639, (Rabbit) 100341296, (Pig) 100626281, (Mouse) 212503, (Rat) 293589, (Guinea pig) 100725437
Molecular Function: DNA binding protein DNA methyltransferase methyltransferase nucleic acid binding oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.