Invitrogen
Manufacturer Code:PA525758
Catalog # PIPA525758
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 467-496 amino acids from the central region of human PAOX |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: exo-N4-amino; PAO peroxisomal N(1)-acetyl-spermine/spermidine oxidase peroxisomal N1-acetyl-spermine/spermidine oxidase; Peroxisomal N(1)-acetyl-spermine/spermidine oxidase; peroxisomal N1-acetyl-spermine/spermidine oxidase; Polyamine oxidase
Gene Aliases: 2410012F02Rik; AI118225; PAO; PAOX; UNQ1923/PRO4398
UniProt ID: (Human) Q6QHF9, (Mouse) Q8C0L6
Entrez Gene ID: (Human) 196743, (Mouse) 212503
Molecular Function: DNA binding protein DNA methyltransferase methyltransferase nucleic acid binding oxidase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.