Novus Biologicals
Manufacturer Code:NBP273244AF647
Catalog # N273244A647
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3C4 |
Immunogen | Full length recombinant protein of human PANK3 (NP_078870) produced in HEK293T cells. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: DKFZp686O22247 EC 2.7.1.33 FLJ12899 hPanK3 MGC16863 pantothenate kinase 3 Pantothenic acid kinase 3; hPanK3; Pantothenate kinase 3; Pantothenic acid kinase 3
Gene Aliases: PANK3
UniProt ID: (Human) Q9H999
Entrez Gene ID: (Human) 79646
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.