Invitrogen
Manufacturer Code:PA513962
Catalog # PIPA513962
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 180-210 amino acids from the central region of human PANK2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hallervorden-Spatz syndrome; hPanK2; pantothenate kinase 2 (Hallervorden-Spatz syndrome); Pantothenate kinase 2 mitochondrial Pantothenic acid kinase 2 hPANK2; Pantothenate kinase 2, mitochondrial; Pantothenic acid kinase 2
Gene Aliases: 4933409I19Rik; AI642621; C20orf48; HARP; HSS; NBIA1; PANK2; PKAN
UniProt ID: (Human) Q9BZ23
Entrez Gene ID: (Human) 80025, (Mouse) 74450
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.