Invitrogen
Manufacturer Code:PA5116411
Catalog # PIPA5116411
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 75-105 amino acids from the N-terminal region of human PANK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPanK; hPanK hPanK1 mPank Pank PANK1 Pank1b Pantothenate kinase 1 pantothenic acid kinase 1; mPank; Pantothenate kinase 1; Pantothenic acid kinase 1
Gene Aliases: PANK; PANK1; Pank1b
UniProt ID: (Human) Q8TE04, (Mouse) Q8K4K6
Entrez Gene ID: (Human) 53354, (Mouse) 75735
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.