Novus Biologicals
Manufacturer Code:NBP304203
Catalog # NB093735
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic Peptide of human PANK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.7.1.33 hPanK hPanK1 MGC24596 PANK PANK1a PANK1b pantothenate kinase pantothenate kinase 1 Pantothenic acid kinase 1; hPanK; Pantothenate kinase 1; Pantothenic acid kinase 1
Gene Aliases: PANK; PANK1
UniProt ID: (Human) Q8TE04
Entrez Gene ID: (Human) 53354
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.