Proteintech
Manufacturer Code:124661AP150UL
Catalog # 50-172-7999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PANK1/2/3 Fusion Protein Ag3140 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hallervorden-Spatz syndrome; hPanK; hPanK2; hPanK3; hPanK3 PANK1/2/3 PANK3 pantothenate kinase 3 Pantothenic acid kinase 3; mPank; mPanK3; Pantothenate kinase 1; pantothenate kinase 2 (Hallervorden-Spatz syndrome); Pantothenate kinase 2, mitochondrial; Pantothenate kinase 3; Pantothenic acid kinase 1; Pantothenic acid kinase 2; Pantothenic acid kinase 3
Gene Aliases: 4933409I19Rik; AI642621; C20orf48; HARP; HSS; NBIA1; PANK; PANK1; Pank1b; PANK2; PANK3; PKAN
UniProt ID: (Human) Q8TE04, (Human) Q9BZ23, (Human) Q9H999, (Mouse) Q8K4K6, (Mouse) Q8R2W9
Entrez Gene ID: (Human) 53354, (Human) 80025, (Human) 79646, (Mouse) 75735, (Rat) 294088, (Rat) 296167, (Mouse) 74450, (Mouse) 211347, (Rat) 360511
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.