Invitrogen
Manufacturer Code:PA531079
Catalog # PIPA531079
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 858 and 1090 of Human PAN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inactive ubiquitin carboxyl-terminal hydrolase 52; PAB-dependent poly(A)-specific ribonuclease subunit 2; PAB1P-dependent poly(A)-nuclease; PAB1P-dependent poly(A)-specific ribonuclease; PABP-dependent poly(A) nuclease 2; PAN deadenylation complex catalytic subunit 2; PAN deadenylation complex subunit 2; PAN2 poly(A) specific ribonuclease subunit homolog; PAN2 polyA specific ribonuclease subunit; PAN2-PAN3 deadenylation complex catalytic subunit PAN2; Poly(A)-nuclease deadenylation complex subunit 2; ubiquitin specific peptidase 52; ubiquitin specific protease 52; USP52 PAB-dependent poly(A)-specific ribonuclease subunit 2 PABP-dependent poly(A) nuclease 2 PAN2 polyA specific ribonuclease subunit inactive ubiquitin carboxyl-terminal hydrolase 52 ubiquitin specific peptidase 52 ubiquitin specific protease 52
Gene Aliases: KIAA0710; PAN2; USP52
UniProt ID: (Human) Q504Q3
Entrez Gene ID: (Human) 9924
Molecular Function:
RNA binding protein
cysteine protease
esterase
exoribonuclease
hydrolase
nuclease
nucleic acid binding
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.