Proteintech
Manufacturer Code:164271AP150UL
Catalog # 50-173-0938
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | PAN2 Fusion Protein Ag9670 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AN2 polyA specific ribonuclease subunit homolog; hPAN2 KIAA0710 PAN2 USP52; Inactive ubiquitin carboxyl-terminal hydrolase 52; inactive ubiquitin carboxyl-terminal hydrolase 52 {ECO:0000255|HAMAP-Rule:MF_03182}; PAB-dependent poly(A)-specific ribonuclease subunit 2; PAB-dependent poly(A)-specific ribonuclease subunit PAN2; PAB-dependent poly(A)-specific ribonuclease subunit PAN2 {ECO:0000255|HAMAP-Rule:MF_03182}; PAB1P-dependent poly(A)-nuclease; PAB1P-dependent poly(A)-nuclease {ECO:0000255|HAMAP-Rule:MF_03182}; PAB1P-dependent poly(A)-specific ribonuclease; PABP-dependent poly(A) nuclease 2; PAN deadenylation complex catalytic subunit 2; PAN deadenylation complex catalytic subunit 2 {ECO:0000255|HAMAP-Rule:MF_03182}; PAN deadenylation complex subunit 2; PAN2 poly(A) specific ribonuclease subunit homolog; PAN2 polyA specific ribonuclease subunit; PAN2 polyA specific ribonuclease subunit homolog; PAN2-PAN3 deadenylation complex catalytic subunit PAN2; Poly(A)-nuclease deadenylation complex subunit 2; ubiquitin specific peptidase 52; ubiquitin specific protease 52
Gene Aliases: 1200014O24Rik; AI047843; AW742773; KIAA0710; mKIAA0710; PAN2; USP52
UniProt ID: (Human) Q504Q3, (Rat) Q6IE70, (Mouse) Q8BGF7
Entrez Gene ID: (Human) 9924, (Rat) 408200, (Mouse) 103135
Molecular Function:
RNA binding protein
cysteine protease
esterase
exoribonuclease
hydrolase
nuclease
nucleic acid binding
protease
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.