Invitrogen
Manufacturer Code:PA544528
Catalog # PIPA544528
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human PALM |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Paralemmin; Paralemmin KIAA0270; Paralemmin-1
Gene Aliases: KIAA0270; PALM
UniProt ID: (Pig) Q2MJV8, (Human) O75781, (Rat) Q920Q0, (Mouse) Q9Z0P4
Entrez Gene ID: (Pig) 654410, (Dog) 485098, (Rabbit) 103347410, (Human) 5064, (Rat) 170673, (Mouse) 18483
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.