Invitrogen
Manufacturer Code:PA569540
Catalog # PIPA569540
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide corresponding to a region of rat PAK4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: p21 (CDKN1A)-activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 4; p21(CDKN1A)-activated kinase 4; p21-activated kinase 4; PAK-4; PAK-4 p21(CDKN1A)-activated kinase 4 p21-activated kinase 4 protein kinase related to S. cerevisiae STE20 effector for Cdc42Hs serine/threonine-protein kinase PAK 4; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; Serine/threonine-protein kinase PAK 4
Gene Aliases: 5730488L07Rik; AW555722; im:5916170; KIAA1142; mKIAA1142; PAK4; wu:fd12b01; zgc:92014
UniProt ID: (Human) O96013, (Mouse) Q8BTW9
Entrez Gene ID: (Dog) 484513, (Rabbit) 100341934, (Bovine) 539742, (Horse) 100067000, (Human) 10298, (Mouse) 70584, (Rat) 292756, (Guinea pig) 100736193, (Zebrafish) 431769
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.