Novus Biologicals
Manufacturer Code:NBP275607
Catalog # NBP275607
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | JB41-84 |
Immunogen | Recombinant protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Rabbit Monoclonal Antibody
Protein Aliases: C-t-PAK2; EC 2.7.11 gamma-PAK p21 (CDKN1A)-activated kinase 2 p21 protein (Cdc42/Rac)-activated kinase 2 p21-activated kinase 2 p58 PAK-2 PAK65EC 2.7.11.1 PAKgamma S6/H4 kinase serine/threonine-protein kinase PAK 2; Gamma-PAK; p21 (CDKN1A)-activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 2; p21-activated kinase 2; p27; p34; p58; PAK-2; PAK-2p27; PAK-2p34; PAK65; S6/H4 kinase; Serine/threonine-protein kinase PAK 2
Gene Aliases: PAK2; PAK65; PAKgamma
UniProt ID: (Human) Q13177
Entrez Gene ID: (Human) 5062
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.