Invitrogen
Manufacturer Code:PA520254
Catalog # PIPA520254
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 14 amino acid peptide from near the amino terminus of human PAK2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C-t-PAK2; Gamma-PAK; p21 (CDKN1A)-activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 2; p21-activated kinase 2; p27; p34; p58; PAK-2; PAK-2p27; PAK-2p34; PAK65; PAK65 PAKgamma PAK-2 S6/H4 kinase gamma-PAK p21 (CDKN1A)-activated kinase 2 p21-activated kinase 2 p58 serine/threonine-protein kinase PAK 2; S6/H4 kinase; Serine/threonine-protein kinase PAK 2
Gene Aliases: 5330420P17Rik; A130002K10Rik; AI836325; D16Ertd269e; gamma-PAK; mKIAA4182; PAK-2; PAK2; PAK65; PAKgamma
UniProt ID: (Human) Q13177, (Mouse) Q8CIN4, (Rat) Q64303
Entrez Gene ID: (Human) 5062, (Mouse) 224105, (Rat) 29432
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.