Invitrogen
Manufacturer Code:PA529224
Catalog # PIPA529224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 167 and 436 of Human PAK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activated protein kinase alpha; Alpha-PAK; CDC42/RAC effector kinase PAK-A; p21 (CDKN1A)-activated kinase 1; p21 protein (Cdc42/Rac)-activated kinase 1; p21-activated kinase 1; p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast); p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related); p65-PAK; p68-PAK; PAK-1; PAKalpha STE20 homolog yeast alpha-PAK p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog yeast) p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related) p65-PAK serine/threonine-protein kinase PAK 1; Protein kinase MUK2; Serine/threonine-protein kinase PAK 1; STE20 homolog, yeast
Gene Aliases: AW045634; PAK-1; PAK1; Paka; PAKalpha
UniProt ID: (Human) Q13153, (Rat) P35465
Entrez Gene ID: (Human) 5058, (Mouse) 18479, (Rat) 29431
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.