Invitrogen
Manufacturer Code:PA517789
Catalog # PIPA517789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues at the carboxy-terminus of human PAK1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: activated protein kinase alpha; adriamycin resistance-associated; Alpha-PAK; Beta-PAK; C-t-PAK2; CDC42/RAC effector kinase PAK-A; CDC42/RAC effector kinase PAK-B; Gamma-PAK; Oligophrenin-3; p21 (CDKN1A)-activated kinase 1; p21 (CDKN1A)-activated kinase 2; p21 (CDKN1A)-activated kinase 3; p21 protein (Cdc42/Rac)-activated kinase 1; p21 protein (Cdc42/Rac)-activated kinase 2; p21 protein (Cdc42/Rac)-activated kinase 3; p21-activated kinase 1; p21-activated kinase 2; p21-activated kinase 3; p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast); p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related); p21/Cdc42/Rac1-activated kinase 3 (yeast Ste20-related); p27; p34; p58; p65-PAK; p68-PAK; PAK-1; PAK-1 STE20 homolog yeast alpha-PAK p21-activated kinase 1 p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog yeast) p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related) p65-PAK serine/threonine-protein kinase PAK 1; PAK-2; PAK-2p27; PAK-2p34; PAK-3; PAK65; Protein kinase MUK2; S6/H4 kinase; Serine/threonine-protein kinase PAK 1; Serine/threonine-protein kinase PAK 2; Serine/threonine-protein kinase PAK 3; STE20 homolog, yeast
Gene Aliases: 5330420P17Rik; A130002K10Rik; AI836325; ARA; AW045634; beta-PAK; bPAK; D16Ertd269e; gamma-PAK; mKIAA4182; MRX30; MRX47; OPHN3; PAK-1; PAK-2; PAK-3; PAK1; PAK2; PAK3; PAK3beta; PAK65; Pak65alpha; Pak65beta; Paka; PAKalpha; Pakb; PAKgamma; Stk4
UniProt ID: (Human) Q13153, (Human) Q13177, (Human) O75914, (Rat) P35465, (Mouse) Q8CIN4, (Rat) Q64303, (Mouse) Q8K1R5, (Rat) Q62829
Entrez Gene ID: (Human) 5058, (Human) 5062, (Human) 5063, (Mouse) 18479, (Guinea pig) 100714662, (Rat) 29431, (Mouse) 224105, (Rat) 29432, (Mouse) 18481, (Rat) 29433
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.