Invitrogen
Manufacturer Code:OSP00019W
Catalog # PIOSP00019W
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Fruit fly |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide from the 4th cytoplasmic loop of drosophila Pain. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CG15860-PA; CG15860-PB; CG15860-PC; pain-PA; pain-PB; pain-PC; RE03641p; Transient receptor potential cation channel protein painless
Gene Aliases: CG15860; CT31987; Dmel\CG15860; Dmel_CG15860; EP2251; Pain
UniProt ID: (Fruit fly) Q9W0Y6
Entrez Gene ID: (Fruit fly) 37985
Molecular Function:
ion channel
transporter
voltage-gated ion channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.