Invitrogen
Manufacturer Code:PA571768
Catalog # PIPA571768
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 95-123 amino acids from the N-terminal region of human PAICS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AIR carboxylase; AIRC; Multifunctional protein ADE2; Multifunctional protein ADE2 Phosphoribosylaminoimidazole-succinocarboxamide synthase SAICAR synthetase Phosphoribosylaminoimidazole carboxylase AIR carboxylase AIRC PAICS ADE2 AIRC PAIS; multifunctional protein ADE2H1; Phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazole succinocarboxamide synthetase; Phosphoribosylaminoimidazole-succinocarboxamide synthase; SAICAR synthetase
Gene Aliases: 2610511I09Rik; ADE2; ADE2H1; AIRC; PAICS; PAIS
UniProt ID: (Human) P22234, (Mouse) Q9DCL9
Entrez Gene ID: (Human) 10606, (Mouse) 67054
Molecular Function: ligase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.