Invitrogen
Manufacturer Code:MA537648
Catalog # PIMA537648
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 717CT21.1.1 |
Immunogen | KLH conjugated synthetic peptide between 127-161 amino acids from the Central region of human PAH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AW106920 PAH PH Phe-4-monooxygenase phenylalanine 4-monooxygenase phenylalanine hydroxylase phenylalanine-4-hydroxylase PKU PKU1; PAH; Phe-4-monooxygenase; phenylalanine 4-monooxygenase; Phenylalanine-4-hydroxylase
Gene Aliases: AW106920; PAH; PH; PKU; PKU1
UniProt ID: (Human) P00439, (Mouse) P16331
Entrez Gene ID: (Human) 5053, (Mouse) 18478, (Rat) 24616
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.