Invitrogen
Manufacturer Code:PA524843
Catalog # PIPA524843
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 132-161 amino acids from the central region of human PAH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: PAH; PH PKU PKU1 phe-4-monooxygenase phenylalanine 4-monooxygenase phenylalanine-4-hydroxylase; Phe-4-monooxygenase; phenylalanine 4-monooxygenase; Phenylalanine-4-hydroxylase
Gene Aliases: AW106920; PAH; PH; PKU; PKU1
UniProt ID: (Human) P00439, (Mouse) P16331
Entrez Gene ID: (Human) 5053, (Mouse) 18478
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.