Invitrogen
Manufacturer Code:PA520828
Catalog # PIPA520828
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 15 amino acid peptide from near the carboxy terminus of human GAS. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GAS; GAS PA1 PAXIP1-associated protein 1 PTIP-associated 1 protein PTIP-associated protein 1 glutamate-rich coactivator interacting with SRC1/NCOA1; glutamate-rich coactivator associated with SRC1; Glutamate-rich coactivator interacting with SRC1; glutamate-rich coactivator interacting with SRC1/NCOA1; PAXIP1 associated glutamate-rich protein 1; PAXIP1-associated glutamate-rich protein 1; PAXIP1-associated glutamate-rich protein 1A; PAXIP1-associated protein 1; PTIP-associated 1; PTIP-associated 1 protein; PTIP-associated protein 1
Gene Aliases: 2900092E17Rik; C16orf53; C77040; GAS; PA1; Pagr; PAGR1; Pagr1a
UniProt ID: (Human) Q9BTK6, (Mouse) Q99L02
Entrez Gene ID: (Human) 79447, (Mouse) 67278
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.