Novus Biologicals
Manufacturer Code:NBP246314
Catalog # NBP246314
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5D2 |
Immunogen | Full length human recombinant protein of human PAFAH1B3 (NP_001139411) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.1.1.47 FLJ44990 PAF acetylhydrolase 29 kDa subunit PAF-AH 29 kDa subunit PAFAH subunit gamma PAF-AH subunit gamma PAF-AH1b alpha 1 subunit PAFAHG platelet-activating factor acetylhydrolase 1b catalytic subunit 3 (29kDa) platelet-activating factor acetylhydrolase IB subunit gamma platelet-activating factor acetylhydrolase isoform Ib gamma subunit (29kD) platelet-activating factor acetylhydrolase isoform Ib gamma subunit 29kDa platelet-activating factor acetylhydrolase isoform Ib subunit 3 (29kDa); PAF acetylhydrolase 29 kDa subunit; PAF-AH 29 kDa subunit; PAF-AH subunit gamma; PAF-AH1b alpha 1 subunit; PAFAH subunit gamma; platelet-activating factor acetylhydrolase 1b, catalytic subunit 3 (29kDa); Platelet-activating factor acetylhydrolase IB subunit alpha1; platelet-activating factor acetylhydrolase, isoform Ib, subunit 3 (29kDa)
Gene Aliases: PAFAH1B3; PAFAHG
UniProt ID: (Human) Q15102
Entrez Gene ID: (Human) 5050
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.